Learn More
ABclonal Technology LDHC Rabbit pAb
SDP
Catalog No. 5000105624
Encompass_Preferred
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5 untranslated region.
Catalog No. 50-001-05624 Supplier ABclonal Technology Supplier No. A15003500UL
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5 untranslated region.