Learn More
Apexbio Technology LLC LDK378 1032900-25-6 200mg
SDP
Catalog No. 5000094467
Encompass_Preferred
LDK378 (CAS 1032900-25-6) is a potent inhibitor of anaplastic lymphoma kinase (ALK) a receptor tyrosine kinase within the insulin receptor superfamily It exhibits an in vitro IC50 of 200 pM for ALK and displays moderate to high inhibitory activity against a limited set of other kinases including IGF-1R (IC50 8 nM) InsR (IC50 7 nM) and STK22D (IC50 23 nM) LDK378 suppresses proliferation of Ba/F3 cells transfected with NPM-ALK fusion and Karpas 299 human lymphoma cells harboring NPM-ALK with IC50 values of 22 8 nM and 26 nM respectively The compound is widely used in preclinical studies to investigate ALK-driven signaling and its potential as a therapeutic target in oncology
Catalog No. 50-000-94467 Supplier Apexbio Technology LLC Supplier No. A8328200
LDK378 (CAS 1032900-25-6) is a potent inhibitor of anaplastic lymphoma kinase (ALK) a receptor tyrosine kinase within the insulin receptor superfamily It exhibits an in vitro IC50 of 200 pM for ALK and displays moderate to high inhibitory activity against a limited set of other kinases including IGF-1R (IC50 8 nM) InsR (IC50 7 nM) and STK22D (IC50 23 nM) LDK378 suppresses proliferation of Ba/F3 cells transfected with NPM-ALK fusion and Karpas 299 human lymphoma cells harboring NPM-ALK with IC50 values of 22 8 nM and 26 nM respectively The compound is widely used in preclinical studies to investigate ALK-driven signaling and its potential as a therapeutic target in oncology