Learn More
Crystal Chem Inc LDL-Cholesterol Assay Kit, 96-tests
SDP
Catalog No. 501947664
Encompass_Preferred
The LDL Cholesterol Assay is used to quantify levels of LDL cholesterol in human serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 4.5 mg/dL. Dynamic range of 4.5 - 245 mg/dL
Catalog No. 50-194-7664 Supplier Crystal Chem Inc Supplier No. 80069
The LDL Cholesterol Assay is used to quantify levels of LDL cholesterol in human serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 4.5 mg/dL. Dynamic range of 4.5 - 245 mg/dL