Learn More
Athens Research And Technology Inc LDL LIPOPROTEINS CELL CULTURE
Supplier:  Athens Research And Technology Inc 1216120412TC5MG
Encompass_Preferred
Lipoproteins, Low Density, HP, Cell Culture: 5 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 2,300,000; Endotoxin Level: <=10 EU/mg. Native; Prepared and purified from fresh, non-frozen human plasma in the Athens Research & Technology laboratory. Lipoproteins, Molecular Biology Reagent
Catalog No. 50-489-869