Learn More
Athens Research And Technology Inc LDL LIPOPROTEINS HU 1MG
Supplier:  Athens Research And Technology Inc 12161204121MG
Encompass_Preferred
Lipoproteins, Low Density, Human Plasma: 1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 2,300,000. Native; Prepared and purified from fresh, non-frozen human plasma in the Athens Research & Tech lab. Lipoproteins, Molecular Biology Reagent
Catalog No. 50-489-865