Learn More
Cayman Chemical LDL RECPTR RabbIt MoncLNa 25
SDP
Supplier:  Cayman Chemical 3807525 TEST
Encompass_Preferred
Immunogen: Recombinant human LDLR • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) LDLR • Application: FC
Catalog No. 50-291-7721