Learn More
Cayman Chemical LDL RECPTR RabbIt MoncLNa
SDP
Catalog No. 502915300
Encompass_Preferred
Immunogen: Recombinant mouse LDLR • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) LDLR • Applications: FC
Catalog No. 50-291-5300 Supplier Cayman Chemical Supplier No. 37077100 TEST
Immunogen: Recombinant mouse LDLR • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) LDLR • Applications: FC