Learn More
Cayman Chemical LDL RECPTR RabbIt MoncLNa
SDP
Catalog No. 502915232
Encompass_Preferred
A peptide orexin receptor 1 agonist selectively induces calcium mobilization in CHO cells overexpressing human OX1R over CHO cells overexpressing human OX2R (EC50s 8.29 and 187 nM respectively)
Catalog No. 50-291-5232 Supplier Cayman Chemical Supplier No. 37076100 UL
A peptide orexin receptor 1 agonist selectively induces calcium mobilization in CHO cells overexpressing human OX1R over CHO cells overexpressing human OX2R (EC50s 8.29 and 187 nM respectively)