Learn More
Cayman Chemical LDL RECPTR RabbIt MoncLNa 50uL
SDP
Catalog No. 502914914
Encompass_Preferred
A phenolic glucoside with in vitro neuroprotective activity reduces glutamate-induced neurotoxicity by 19.3 in PC12 cells at 10 UM
Catalog No. 50-291-4914 Supplier Cayman Chemical Supplier No. 3707650 UL
A phenolic glucoside with in vitro neuroprotective activity reduces glutamate-induced neurotoxicity by 19.3 in PC12 cells at 10 UM