Learn More
Cayman Chemical LDL Uptake Flow Cytometry48tSt
SDP
Catalog No. 502892303
Encompass_Preferred
Cayman’s LDL Uptake Flow Cytometry Assay Kit employs human LDL conjugated to DyLight™ 488 as a convenient tool for studying the uptake of LDL in cultured cells. Flow cytometry provides the advantage of assessing the uptake of LDL at the single-cell level. In addition, multiplexing with other markers, such as LDLR expression, is possible to gain more information from a single experiment. Lovastatin is included in this kit as a control modulator of LDL uptake by hepatocytes. The reagents provided in this kit are sufficient to test 48 samples by FC.
Catalog No. 50-289-2303 Supplier Cayman Chemical Supplier No. 60147048 TEST
Cayman’s LDL Uptake Flow Cytometry Assay Kit employs human LDL conjugated to DyLight™ 488 as a convenient tool for studying the uptake of LDL in cultured cells. Flow cytometry provides the advantage of assessing the uptake of LDL at the single-cell level. In addition, multiplexing with other markers, such as LDLR expression, is possible to gain more information from a single experiment. Lovastatin is included in this kit as a control modulator of LDL uptake by hepatocytes. The reagents provided in this kit are sufficient to test 48 samples by FC.