Learn More
Cell Biolabs Inc LDL/VLDL and HDL Purification Kit (Ultracentifugation-Free), 10 preps
Catalog No. 50673650
Encompass_Preferred
Cell Biolabs' LDL/VLDL and HDL Purification Kit uses dextran sulfate to selectively and separately purify LDL/VLDL and HDL fractions for various downstream applications. No ultracentrifuge is necessary. Each kit provides sufficient reagents for 10 preps.
Catalog No. 50-673-650 Supplier Cell Biolabs Inc Supplier No. STA608
Cell Biolabs' LDL/VLDL and HDL Purification Kit uses dextran sulfate to selectively and separately purify LDL/VLDL and HDL fractions for various downstream applications. No ultracentrifuge is necessary. Each kit provides sufficient reagents for 10 preps.