Learn More
ACROBiosystems ACROBiosystems Human LDL R Protein, His Tag
Catalog No. 501956573
Encompass_Preferred
Recombinant Protein;100UG;Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession # NP_000518.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.9 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PCSK9 (D374Y), Avitag,His Tag (Cat. No. PCY-H82E7) with a linear range of 4-63 ng/mL (QC tested).
Catalog No. 50-195-6573 Supplier ACROBiosystems Supplier No. LDRH5224100UG
Recombinant Protein;100UG;Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession # NP_000518.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.9 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PCSK9 (D374Y), Avitag,His Tag (Cat. No. PCY-H82E7) with a linear range of 4-63 ng/mL (QC tested).