Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LDLR ELISA Kit

Catalog No. EHLDLRX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHLDLRX5 5 x 96 Tests
EHLDLR 96 Tests
EHLDLRX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHLDLRX5 Supplier Invitrogen™ Supplier No. EHLDLRX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human LDLR ELISA quantitates Hu LDLR in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu LDLR. Principle of the method The Human LDLR solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01130
Assay Range 8.19-2000 pg/mL
Assay Sensitivity 8 pg/mL
Calibration Range 8 pg/mL to 2000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human LDLR. Other species not determined yet.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3; FH; FHC; LDLCQ2
Gene ID (Entrez) 3949
Gene Symbol LDLR
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases LDL receptor
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Virology
Sample Volume Plasma 2.5 μL, Serum 2.5 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym FH, FHC, LDLCQ2
Target LDLR
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.