Learn More
Sino Biological LDLR / LDL Receptor Antibody (FITC), Rabbit MAb 100Tests
Supplier:  Sino Biological 50305R032F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR / LDL Receptor (rM LDLR / LDL Receptor; Catalog#50305-M08H; NP_034830.2; Met 1-Arg 790) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-224-8254