Learn More
Sino Biological LDLR / LDL Receptor Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 50305R032100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR / LDL Receptor (rM LDLR; Catalog#50305-M08H; Met 1-Arg 790; NP_034830.2).
Catalog No. 50-224-8248