Learn More
Aviva Systems Biology Corp LDLRAD3 Antibody - middle region (ARP53301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LDLRAD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LDLRAD3. Alternate names: LRAD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-824