Learn More
ACROBiosystems Recombinant Protein;Mouse LDL R Protein;HEK293;100UG;LDR,LDLR,FH,FHC,LDLCQ2,LDL Receptor
Catalog No. 501658706
Encompass_Preferred
Recombinant Protein;Mouse LDL R Protein;100UG;Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession # NP_034830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 86.2 kDa. The protein migrates as 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PCSK9, Strep Tag (Cat. No. PC9-H528c) at 10 μg/mL (100 μL/well) can bind Mouse LDL R, His Tag (Cat. No. LDR-M52H8) with a linear range of 0.102-1.6 μg/mL (QC tested).
Catalog No. 50-165-8706 Supplier ACROBiosystems Supplier No. LDRM52H8100UG
Recombinant Protein;Mouse LDL R Protein;100UG;Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession # NP_034830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 86.2 kDa. The protein migrates as 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PCSK9, Strep Tag (Cat. No. PC9-H528c) at 10 μg/mL (100 μL/well) can bind Mouse LDL R, His Tag (Cat. No. LDR-M52H8) with a linear range of 0.102-1.6 μg/mL (QC tested).