Learn More
ACROBiosystems Recombinant Protein;Cynomolgus LDL R Protein;HEK293;1MG;LDR,LDLR,FH,FHC,LDLCQ2,LDL Receptor
Catalog No. 501658433
Encompass_Preferred
Recombinant Protein;Cynomolgus LDL R Protein;1MG;Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession # XP_005588053.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 86.8 kDa. The protein migrates as 135-155 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LDL R, His Tag (Cat. No. LDR-C5222) at 2μg/mL (100 μL/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 0.007-0.256 μg/mL (QC tested).;Immobilized Cynomolgus PCSK9, His Tag (Cat. No. PC9-C5223) at 10μg/mL (100 μL/well) can bind Cynomolgus LDL R, His Tag (Cat. No. LDR-C5222) with a linear range of 0.04-0.256 μg/mL (Routinely tested).
Catalog No. 50-165-8433 Supplier ACROBiosystems Supplier No. LDRC52221MG
Recombinant Protein;Cynomolgus LDL R Protein;1MG;Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession # XP_005588053.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 86.8 kDa. The protein migrates as 135-155 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LDL R, His Tag (Cat. No. LDR-C5222) at 2μg/mL (100 μL/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 0.007-0.256 μg/mL (QC tested).;Immobilized Cynomolgus PCSK9, His Tag (Cat. No. PC9-C5223) at 10μg/mL (100 μL/well) can bind Cynomolgus LDL R, His Tag (Cat. No. LDR-C5222) with a linear range of 0.04-0.256 μg/mL (Routinely tested).