Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LDL R Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;LDR,LDLR,FH,FHC,LDLCQ2,LDL Receptor
Catalog No. 501657768
Encompass_Preferred
Recombinant Protein;Biotinylated Human LDL R Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LDL R, His Tag, Avi Tag (LDR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession # P01130-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.5 kDa. The protein migrates as 90-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PCSK9, His Tag (Cat. No. PC9-H5223) at 10 μg/mL (100 μL/well) can bind Biotinylated Human LDL R, His Tag, Avi Tag (Cat. No. LDR-H82E7) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-165-7768 Supplier ACROBiosystems Supplier No. LDRH82E725UG
Recombinant Protein;Biotinylated Human LDL R Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LDL R, His Tag, Avi Tag (LDR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession # P01130-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.5 kDa. The protein migrates as 90-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PCSK9, His Tag (Cat. No. PC9-H5223) at 10 μg/mL (100 μL/well) can bind Biotinylated Human LDL R, His Tag, Avi Tag (Cat. No. LDR-H82E7) with a linear range of 0.02-0.625 μg/mL (QC tested).