Learn More
WASP LDM Clay Kit Extrusion System 3.0
SCI_ED
Catalog No. S4131948
Small Parts - Not recommended for children under 3 years old.
Catalog No. S4131948 Supplier WASP Supplier No. WU7089
Small Parts - Not recommended for children under 3 years old.
The LDM Clay Kit Extrusion System 3.0 can be used to turn your FDM printer into a hybrid printer with LDM printing capabilities.
  • Air Compressor is necessary for WASP LDM Printing but not included in Clay Kit Add On.
  • Compatible with all air compressors with a minimum of 8 bar.
Standard Configuration includes:
  • LDM Extruder
  • 1.2 mm and 2 mm diameter nozzles
  • 3 L Tank
  • Tank Stand
  • WASP Gauge
  • Teflon Tubing
  • Nylon Tubing
  • 25 lb. Porcelain
  • Manual
  • SD Card
Compatible with the following printers:
  • WASP 2040 Pro
  • WASP 2040 Clay
  • WASP 4070 Pro
  • WASP 40100 Clay
  • WASP 60100 Pro
Specifications
For Use With (Equipment) WASP 2040 Pro, WASP 2040 Clay, WASP 4070 Pro, WASP 40100 Clay, WASP 60100 Pro
Product Type LDM Extruder Kit