Learn More
Selleck Chemical LLC LDN-192960 S3406-5mg
SDP
Catalog No. 503109271
Encompass_Preferred
LDN-192960 (Compd 1) is a potent inhibitor of Haspin and Dual-specificity Tyrosine-regulated Kinase 2 (DYRK2) with IC50 of 10 nM and 48 nM respectively
Catalog No. 50-310-9271 Supplier Selleck Chemical LLC Supplier No. S34065MG
LDN-192960 (Compd 1) is a potent inhibitor of Haspin and Dual-specificity Tyrosine-regulated Kinase 2 (DYRK2) with IC50 of 10 nM and 48 nM respectively