Learn More
TARGETMOL CHEMICALS INC LDN-192960 1MG
SDP
Catalog No. 502877295
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. LDN-192960 is a potent inhibitor of Haspin and DYRK2 (IC50s 10 nM and 48 nM).
Catalog No. 50-287-7295 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ01111MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. LDN-192960 is a potent inhibitor of Haspin and DYRK2 (IC50s 10 nM and 48 nM).