Learn More
Medchemexpress LLC HY-13455A 5mg Medchemexpress, LDN-192960 (hydrochloride) CAS:2309172-48-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13455A5MG
Encompass_Preferred
Medchemexpress, HY-13455A 5mg LDN-192960 (HCl) CAS:2309172-48-1 LDN-192960 hydrochloride is an inhibitor of Haspin and Dual-specificity Tyrosine-regulated Kinase 2 (DYRK2) with IC50s of 10 nM and 48 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9853