Learn More
Cayman Chemical LDN-192960 (hydrochloride), Purity: ≥98%, CAS: 2309172-48-1, 50 mg
SDP
Catalog No. 502983684
Encompass_Preferred
An inhibitor of haspin and DYRK2 (IC50s = 10 and 48 nM, respectively); selective for haspin and DYRK2 over TRKB, CLK1, DYRK1A, DYRK3, ROS, HIPK1, HIPK2, PIM-1, and PIM-2 kinases (IC50s = 720-91,000 nM)
Catalog No. 50-298-3684 Supplier Cayman Chemical Supplier No. 1908950 MG
An inhibitor of haspin and DYRK2 (IC50s = 10 and 48 nM, respectively); selective for haspin and DYRK2 over TRKB, CLK1, DYRK1A, DYRK3, ROS, HIPK1, HIPK2, PIM-1, and PIM-2 kinases (IC50s = 720-91,000 nM)