Learn More
Selleck Chemical LLC LDN-193189 2HCl
SDP
Catalog No. 5000003653
Encompass_Preferred
LDN-193189 (DM3189) 2HCl is a selective BMP signaling inhibitor inhibits the ALK1 ALK2 ALK3 and ALK6 with IC50s of 0 8 nM 0 8 nM 5 3 nM and 16 7 nM in the kinase assay respectively LDN-193189 inhibits the transcriptional activity of the BMP type I receptors ALK2 and ALK3 with IC50s of 5 nM and 30 nM in C2C12 cells respectively exhibits 200-fold selectivity for BMP versus TGF-
Catalog No. 50-000-03653 Supplier Selleck Chemical LLC Supplier No. S750750MG
LDN-193189 (DM3189) 2HCl is a selective BMP signaling inhibitor inhibits the ALK1 ALK2 ALK3 and ALK6 with IC50s of 0 8 nM 0 8 nM 5 3 nM and 16 7 nM in the kinase assay respectively LDN-193189 inhibits the transcriptional activity of the BMP type I receptors ALK2 and ALK3 with IC50s of 5 nM and 30 nM in C2C12 cells respectively exhibits 200-fold selectivity for BMP versus TGF-