Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ldn-193189 (dihydrochloride) | 1435934-00-1 | 99.8% | 25 MG
SDP

Catalog No. 5000230321
Encompass_Preferred

LDN-193189 dihydrochloride is a potent, selective small-molecule inhibitor of BMP type I receptors (ALK2 and ALK3). It blocks BMP-mediated SMAD1/5/8 signaling and is used in biochemical and cell-based research to probe BMP pathway biology.

  • Potent ALK2 inhibitor (IC50 = 5 nM) and ALK3 inhibitor (IC50 = 30 nM).
  • Selective inhibition of BMP type I receptor-mediated SMAD1/5/8 phosphorylation.
  • Molecular weight 479.40 Da and formula C25H24Cl2N6.
  • High purity (approximately 99.8%).
  • Available in small research-scale quantities suitable for in vitro assays.

Catalog No. 50-002-30321 Supplier Medchemexpress LLC Supplier No. HY12071B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-193189 dihydrochloride is a potent, selective small-molecule inhibitor of BMP type I receptors (ALK2 and ALK3). It blocks BMP-mediated SMAD1/5/8 signaling and is used in biochemical and cell-based research to probe BMP pathway biology.

  • Potent ALK2 inhibitor (IC50 = 5 nM) and ALK3 inhibitor (IC50 = 30 nM).
  • Selective inhibition of BMP type I receptor-mediated SMAD1/5/8 phosphorylation.
  • Molecular weight 479.40 Da and formula C25H24Cl2N6.
  • High purity (approximately 99.8%).
  • Available in small research-scale quantities suitable for in vitro assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.