Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ldn-193189 dihydrochloride | 1435934-00-1 | 99.8% | 5 MG
SDP

Catalog No. 5000233844
Encompass_Preferred

LDN-193189 dihydrochloride is a potent small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors, primarily ALK2 and ALK3, used in biochemical and cell-based research to modulate BMP signaling and study related disease models such as fibrodysplasia ossificans progressiva.

  • Potent inhibitor of BMP type I receptors (ALK2, ALK3).
  • High purity suitable for research applications.
  • Molecular formula C25H24Cl2N6; molecular weight 479.40 g/mol.
  • Solid, orange to red appearance for easy identification.
  • Suitable for cell-based and biochemical assays.

Catalog No. 50-002-33844 Supplier Medchemexpress LLC Supplier No. HY12071B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-193189 dihydrochloride is a potent small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors, primarily ALK2 and ALK3, used in biochemical and cell-based research to modulate BMP signaling and study related disease models such as fibrodysplasia ossificans progressiva.

  • Potent inhibitor of BMP type I receptors (ALK2, ALK3).
  • High purity suitable for research applications.
  • Molecular formula C25H24Cl2N6; molecular weight 479.40 g/mol.
  • Solid, orange to red appearance for easy identification.
  • Suitable for cell-based and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.