Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LDN-193189 monohydrochloride | 1062368-62-0 | >=98.0% | 25 MG
SDP

Catalog No. 5000230112
Encompass_Preferred

LDN193189 hydrochloride is the hydrochloride salt of LDN193189, a selective small-molecule inhibitor of BMP type I receptors used as a biochemical reagent for in vitro research and biochemical assays.

  • Hydrochloride salt form for improved solubility.
  • Molecular formula C25H23ClN6.
  • Molecular weight 442.94 g/mol.
  • CAS number 1062368-62-0.
  • Typical purity ≥98.0% (HPLC).
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or diagnostic applications.

Catalog No. 50-002-30112 Supplier Medchemexpress LLC Supplier No. HY12071C25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN193189 hydrochloride is the hydrochloride salt of LDN193189, a selective small-molecule inhibitor of BMP type I receptors used as a biochemical reagent for in vitro research and biochemical assays.

  • Hydrochloride salt form for improved solubility.
  • Molecular formula C25H23ClN6.
  • Molecular weight 442.94 g/mol.
  • CAS number 1062368-62-0.
  • Typical purity ≥98.0% (HPLC).
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or diagnostic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.