Learn More
Apexbio Technology LLC LDN 209929 dihydrochloride 10mg
SDP
Catalog No. 5000087676
Encompass_Preferred
LDN 209929 dihydrochloride is a small-molecule inhibitor targeting haspin kinase a serine/threonine kinase It is designed to inhibit haspin kinase activity thereby regulating chromosomal alignment and mitotic progression LDN 209929 dihydrochloride exerts its biological activity primarily through selective inhibition of haspin kinase It demonstrates an IC50 of approximately 55 nM and exhibits around 180-fold selectivity over DYRK2 kinase Based on these pharmacological properties LDN 209929 dihydrochloride holds research potential in oncology studies related to aberrant mitotic control and tumor cell proliferation
Catalog No. 50-000-87676 Supplier Apexbio Technology LLC Supplier No. B778510
Add to Cart
Add to Cart
LDN 209929 dihydrochloride is a small-molecule inhibitor targeting haspin kinase a serine/threonine kinase It is designed to inhibit haspin kinase activity thereby regulating chromosomal alignment and mitotic progression LDN 209929 dihydrochloride exerts its biological activity primarily through selective inhibition of haspin kinase It demonstrates an IC50 of approximately 55 nM and exhibits around 180-fold selectivity over DYRK2 kinase Based on these pharmacological properties LDN 209929 dihydrochloride holds research potential in oncology studies related to aberrant mitotic control and tumor cell proliferation