Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LDN-209929 dihydrochloride | 1784281-97-5 | MFCD28133376 | 98.3% | 405.77 g/mol | C17H19Cl3N2OS | 5 MG
SDP

Catalog No. 5000263336
Encompass_Preferred

LDN-209929 dihydrochloride is the dihydrochloride salt of a potent, selective small-molecule inhibitor of haspin kinase (reported IC50 = 55 nM) with approximately 180-fold selectivity over DYRK2. Supplied as a yellow-orange solid for biochemical and cellular research, it is commonly provided in small research pack sizes and is typically ≥98% pure.

  • Potent haspin kinase inhibition (IC50 = 55 nM).
  • Approximately 180-fold selectivity versus DYRK2.
  • Dihydrochloride salt, solid, yellow to orange.
  • Molecular weight 405.77 g/mol.
  • Purity approximately 98.3% by HPLC.
  • Storage: 4°C sealed; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in small research sizes including 5 mg.

Catalog No. 50-002-63336 Supplier Medchemexpress LLC Supplier No. HY1103205MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-209929 dihydrochloride is the dihydrochloride salt of a potent, selective small-molecule inhibitor of haspin kinase (reported IC50 = 55 nM) with approximately 180-fold selectivity over DYRK2. Supplied as a yellow-orange solid for biochemical and cellular research, it is commonly provided in small research pack sizes and is typically ≥98% pure.

  • Potent haspin kinase inhibition (IC50 = 55 nM).
  • Approximately 180-fold selectivity versus DYRK2.
  • Dihydrochloride salt, solid, yellow to orange.
  • Molecular weight 405.77 g/mol.
  • Purity approximately 98.3% by HPLC.
  • Storage: 4°C sealed; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in small research sizes including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.