Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ldn-211904 oxalate | 1198408-78-4 | MFCD28023573 | 99.0% | 444.87 g·mol⁻¹ | C21H21ClN4O5 | 1 ML
SDP

Catalog No. 5000241054
Encompass_Preferred

LDN-211904 oxalate is the oxalate salt of a potent, reversible EphB3 kinase inhibitor supplied for in vitro biochemical and cellular research. Available as solid material and as a ready-to-use DMSO stock solution, it includes manufacturer guidance for solubility and storage to support assay development.

  • Potent and reversible EphB3 inhibitor with low-nanomolar activity.
  • High chemical purity: 99.0%.
  • Molecular weight 444.87 g·mol⁻1 and formula C21H21ClN4O5.
  • Available as solid (1 mg, 5 mg, 10 mg) and prepared DMSO stock (10 mM in 1 mL).
  • Soluble ≥ 2.5 mg/mL (approximately 5.62 mM) in 10% DMSO and compatible vehicles.
  • Storage guidance: -20°C sealed; in solvent -80°C up to 6 months.

Catalog No. 50-002-41054 Supplier Medchemexpress LLC Supplier No. HY10746010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-211904 oxalate is the oxalate salt of a potent, reversible EphB3 kinase inhibitor supplied for in vitro biochemical and cellular research. Available as solid material and as a ready-to-use DMSO stock solution, it includes manufacturer guidance for solubility and storage to support assay development.

  • Potent and reversible EphB3 inhibitor with low-nanomolar activity.
  • High chemical purity: 99.0%.
  • Molecular weight 444.87 g·mol⁻1 and formula C21H21ClN4O5.
  • Available as solid (1 mg, 5 mg, 10 mg) and prepared DMSO stock (10 mM in 1 mL).
  • Soluble ≥ 2.5 mg/mL (approximately 5.62 mM) in 10% DMSO and compatible vehicles.
  • Storage guidance: -20°C sealed; in solvent -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.