Learn More
Medchemexpress LLC Ldn-212320 | 894002-50-7 | 98.9% | 10 MM 1 ML
SDP
Catalog No. 5000338461
Encompass_Preferred
LDN-212320 is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator, acting at the translational level. It prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC.
  • Glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator
  • Acts at the translational level
  • Prevents nociceptive pain
  • Upregulates astroglial GLT-1 expression in the hippocampus and ACC
Catalog No. 50-003-38461 Supplier Medchemexpress LLC Supplier No. HY1274110MM/1ML
LDN-212320 is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator, acting at the translational level. It prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC.
  • Glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator
  • Acts at the translational level
  • Prevents nociceptive pain
  • Upregulates astroglial GLT-1 expression in the hippocampus and ACC