Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LDN-212320 | 894002-50-7 | 98.92% | 100 MG
SDP

Catalog No. 5000342800
Encompass_Preferred

LDN-212320 is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator at the translational level. It prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC. This compound is intended for research use only and is not sold to patients.

  • Glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator
  • Functions at the translational level
  • Prevents nociceptive pain by upregulating astroglial GLT-1 expression
  • Intended for research use only
  • Molecular weight: 293.39
  • Molecular formula: C17H15N3S
  • Target: EAAT2
  • Pathway: Membrane transporter/ion channel

Catalog No. 50-003-42800 Supplier Medchemexpress LLC Supplier No. HY12741100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-212320 is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator at the translational level. It prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC. This compound is intended for research use only and is not sold to patients.

  • Glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator
  • Functions at the translational level
  • Prevents nociceptive pain by upregulating astroglial GLT-1 expression
  • Intended for research use only
  • Molecular weight: 293.39
  • Molecular formula: C17H15N3S
  • Target: EAAT2
  • Pathway: Membrane transporter/ion channel

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.