Learn More
TARGETMOL CHEMICALS INC LDN-212854 10MG
SDP
Catalog No. 502581060
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LDN-212854 (BMP Inhibitor III) a novel BMP inhibitor exhibits greater selectivity for BMP versus the TGF-(beta) type I receptors; possesses differences towards ALK2(IC50=1.3 nM) versus ALK1/3 compared to other inhibitors. purity: 99%
Catalog No. 50-258-1060 Supplier TARGETMOL CHEMICALS INC Supplier No. T190010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. LDN-212854 (BMP Inhibitor III) a novel BMP inhibitor exhibits greater selectivity for BMP versus the TGF-(beta) type I receptors; possesses differences towards ALK2(IC50=1.3 nM) versus ALK1/3 compared to other inhibitors. purity: 99%