Learn More
Selleck Chemical LLC LDN-212854
SDP
Catalog No. 5000002125
Encompass_Preferred
LDN-212854 (BMP Inhibitor III) is a potent and selective BMP receptor inhibitor with IC50 of 1 3 nM for ALK2 about 2- 66- 1641- and 7135-fold selectivity over ALK1 ALK3 ALK4 and ALK5 respectively
Catalog No. 50-000-02125 Supplier Selleck Chemical LLC Supplier No. S714750MG
Add to Cart
Add to Cart
LDN-212854 (BMP Inhibitor III) is a potent and selective BMP receptor inhibitor with IC50 of 1 3 nM for ALK2 about 2- 66- 1641- and 7135-fold selectivity over ALK1 ALK3 ALK4 and ALK5 respectively