Learn More
Selleck Chemical LLC LDN-214117
SDP
Supplier:  Selleck Chemical LLC S76271G
Encompass_Preferred
LDN-214117 is a potent and selective BMP type I receptor kinase ALK2 inhibitor with IC50 of 24 nM
Catalog No. 50-000-00240