Learn More
Selleck Chemical LLC LDN-214117
SDP
Catalog No. 5000000255
Encompass_Preferred
LDN-214117 is a potent and selective BMP type I receptor kinase ALK2 inhibitor with IC50 of 24 nM
Catalog No. 50-000-00255 Supplier Selleck Chemical LLC Supplier No. S762710MM/1ML
LDN-214117 is a potent and selective BMP type I receptor kinase ALK2 inhibitor with IC50 of 24 nM