Learn More
TARGETMOL CHEMICALS INC LDN-27219 10MG
SDP
Catalog No. 502577443
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LDN-27219 is a potent hTGase inhibitor with IC50 of 0.6 uM. purity: 99%
Catalog No. 50-257-7443 Supplier TARGETMOL CHEMICALS INC Supplier No. T1183110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LDN-27219 is a potent hTGase inhibitor with IC50 of 0.6 uM. purity: 99%