Learn More
Apexbio Technology LLC LDN 57444, 10mg. Cas: 668467-91-2 MFCD: MFCD09037562. UCH-L1 inhibitor,reversible competitve
SDP
Supplier:  Apexbio Technology LLC A400310
Encompass_Preferred
LDN 57444 is a reversible competitive inhibitor of ubiquitin C-terminal hydrolase L1 (Uch-L1) with IC50 value of 0.88M [1].Besides Uch-L1, LDN 57444 also inhibits Uch-L3 with a higher IC50 value of 25M. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4850