Learn More
Medchemexpress LLC HY-18637 10mg Medchemexpress, LDN-57444 CAS:668467-91-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1863710MG
Encompass_Preferred
Medchemexpress, HY-18637 10mg LDN-57444 CAS:668467-91-2 LDN-57444 is a reversible, competitive and site-directed inhibitor of ubiquitin C-terminal hydrolase L1 (UCH-L1), with an IC50 of 0.88 μM and a Ki of 0.40 μM; LDN-57444 also suppresses UCH-L3 activity, with an IC50 of 25 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0826