Learn More
Selleck Chemical LLC LDN-57444
SDP
Catalog No. 502173257
Encompass_Preferred
LDN-57444 is a reversible competitive proteasome inhibitor for Uch-L1 with IC50 of 0 88 M 28-fold selectivity over isoform Uch-L3
Catalog No. 50-217-3257 Supplier Selleck Chemical LLC Supplier No. S713525MG
LDN-57444 is a reversible competitive proteasome inhibitor for Uch-L1 with IC50 of 0 88 M 28-fold selectivity over isoform Uch-L3