Learn More
Abcam LDN-57444, UCH-L1 inhibitor, 5MG
Catalog No. 502706573
Encompass_Preferred
MW 397.6 Da, Purity >98%. Reversible, competitive ubiquitin C-terminal hydrolase-L1 (UCH-L1) inhibitor (Ki = 0.4 μM. Increases ubiquitinated protein levels and decreases proteasome activity. Also induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6573 Supplier Abcam Supplier No. AB1414875MG
MW 397.6 Da, Purity >98%. Reversible, competitive ubiquitin C-terminal hydrolase-L1 (UCH-L1) inhibitor (Ki = 0.4 μM. Increases ubiquitinated protein levels and decreases proteasome activity. Also induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement