Learn More
Medchemexpress LLC HY-12071 10mg Medchemexpress, LDN193189 CAS:1062368-24-4 Purity:>98%
SDP
Catalog No. 502028850
Encompass_Preferred
Medchemexpress, HY-12071 10mg LDN193189 CAS:1062368-24-4 LDN193189 is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8850 Supplier Medchemexpress LLC Supplier No. HY1207110MG
Medchemexpress, HY-12071 10mg LDN193189 CAS:1062368-24-4 LDN193189 is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.