Learn More
BIOGEMS INTERNATIONAL INC LDN193189 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 10624431MG
Encompass
LDN193189 is a dorsomorphin derivative and potent BMP pathway inhibitor. It is reported to inhibit bone morphogenetic protein (BMP) type I receptors ALK1, ALK2, ALK3, and ALK6. LDN193189 is useful for the promotion of specific cell differentiation and the study of ectopic ossification.
Catalog No. 50-152-9579