Learn More
STEMCELL Technologies LDN193189 (Dihydrochloride), Size: 50 mg
SDP
Supplier:  STEMCELL Technologies 72149
Encompass
Inhibit ALK1, ALK2, ALK3, and ALK6 with LDN193189 (Sanvitale et al. PLoS One, 2013). This bone morphogenetic (BMP) pathway inhibitor promotes the differentiation of human pluripotent stem cells to neural progenitor cells (Chambers et al. Nat Biotechnol, 2012; Kriks et al. Nature, 2011) and neural crest cells (Kreitzer et al. Am J Stem Cells, 2013). LDN193189 is used in cancer research to prevent ovarian cancer cell proliferation (Tsai et al. Cell Rep, 2012) and inhibit prostate and breast cancer tumor growth (Balboni et al. Cancer Res, 2013; Lee et al. Neuropsychopharmacology, 2016). It also promotes the differentiation of sensory epithelial cells of the inner ear from mouse embryonic stem cells (Koehler et al. Nature, 2013). LDN193189 is supplied as the dihydrochloride hydrate form of the molecule with > 98% purity; Molecular weight 479.4 g/mol; CAS Number 1435934-00-1. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5886