Learn More
Apexbio Technology LLC LDN193189 Hydrochloride, 1062368-62-0, MFCD20926415, 5 mg
SDP
Supplier:  Apexbio Technology LLC A35455
Encompass_Preferred
MF: C25H26Cl4N6, MW: 552.33, Solubility: 16.4 mg/mL in DMSO, 27.6 mg/mL in H2O,insoluble in EtOH, Synonyms: LDN 193189 hydrochloride; LDN-193189 hydrochloride. LDN193189 is a selective transcriptional activity morphogenetic protein (BMP) type I receptors inhibitor. It inhibits activin receptor-like kinase-2 (ALK2) and ALK3 with IC50 values of 5 nM and 30 nM, respectively.
Catalog No. 50-101-4595