Learn More
Medchemexpress LLC HY-12071A 5mg , LDN193189 (Tetrahydrochloride) CAS:2310134-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12071A5MG
Encompass_Preferred
HY-12071A 5mg LDN193189 (Tetrahydrochloride) CAS: 2310134-98-4 LDN193189 Tetrahydrochloride is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0598