Learn More
Medchemexpress LLC LDN193189 (tetrahydrochloride) | 2310134-98-4 | 99.8% | 200 MG
SDP
Catalog No. 5000234873
Encompass_Preferred
LDN193189 (tetrahydrochloride) is a small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors that selectively inhibits ALK2 and ALK3 (reported IC50 ≈ 5 nM and 30 nM). It is supplied for research use only. CAS 2310134-98-4; molecular formula C25H26Cl4N6; molecular weight 552.33 g/mol.
  • Selective inhibitor of BMP type I receptors.
  • Potent inhibition of ALK2 and ALK3 at nanomolar concentrations.
  • High reported purity (99.8%).
  • Intended for research use only, not for human therapeutic use.
  • Storage recommendations: sealed at 4°C; in solvent, -80°C (1 year) or -20°C (6 months).
Catalog No. 50-002-34873 Supplier Medchemexpress LLC Supplier No. HY12071A200MG
LDN193189 (tetrahydrochloride) is a small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors that selectively inhibits ALK2 and ALK3 (reported IC50 ≈ 5 nM and 30 nM). It is supplied for research use only. CAS 2310134-98-4; molecular formula C25H26Cl4N6; molecular weight 552.33 g/mol.
  • Selective inhibitor of BMP type I receptors.
  • Potent inhibition of ALK2 and ALK3 at nanomolar concentrations.
  • High reported purity (99.8%).
  • Intended for research use only, not for human therapeutic use.
  • Storage recommendations: sealed at 4°C; in solvent, -80°C (1 year) or -20°C (6 months).