Learn More
Hach Company LDO Calibration Cap
Catalog No. 5000207141 Shop All Hach Company Products
Encompass
LDO Calibration Cap-HQD SeriesSoaker bottle for LDO101 sensor calibration perfectly fits onto LDO101 sensor body It contains sponge its handy design makes it ideal to perform water-saturated air (100%) calibration procedure in laboratory environment While calibrating it is important for accurate results to ensure having the sensor surface temperature and Thermistor measured temperature to be same thus rendering this accessory not appropriate for field use
Catalog No. 50-002-07141 Supplier Hach Company Supplier No. 5811200PC
LDO Calibration Cap-HQD SeriesSoaker bottle for LDO101 sensor calibration perfectly fits onto LDO101 sensor body It contains sponge its handy design makes it ideal to perform water-saturated air (100%) calibration procedure in laboratory environment While calibrating it is important for accurate results to ensure having the sensor surface temperature and Thermistor measured temperature to be same thus rendering this accessory not appropriate for field use