Learn More
Hach Company LDO Model 2 -DO Probe
Catalog No. 5000208098 Shop All Hach Company Products
Encompass
Hach LDO Model 2 -DO Probe with Luminescent Dissolved Oxygen Technology-for Class 1 Division 2Includes LDO probe-calibation bags-and manual
Catalog No. 50-002-08098 Supplier Hach Company Supplier No. 9020000C1D2
Hach LDO Model 2 -DO Probe with Luminescent Dissolved Oxygen Technology-for Class 1 Division 2Includes LDO probe-calibation bags-and manual